MyVolt™ Touch Power Supply
Supplier: VWR International
470234-462
470234-462EA 799 USD
470234-462
MyVolt™ Touch Power Supply
Power Supplies
Ideal choice for protein or nucleic acid separations.
  • Touchscreen control
  • Dual orientation, auto-rotating display
  • Intuitive programming
  • Voltage range 10 to 300 V
  • Amperage range 5 to 400 mA
  • 2 year warranty
The myVolt Touch is a dependable electrophoresis power supply that includes all of the capabilities and specifications needed for efficient separation.