3B Scientific® Inverting Spectacles
Supplier: American 3B Scientific
1000895
470233-768EA 671.99 USD
470233-768
3B Scientific® Inverting Spectacles
Educational Materials Physics Educational Materials Optics Learning Activities
Inverting spectacles complete with two fully rotating inverting prisms in a shielded spectacle frame.
  • Demonstration of the path of rays through an inverting prism
  • Drawing and writing
  • Seeing, picking up and balancing objects
  • Throwing and catching a ball
  • Pouring water
Inverting Spectacles Complete with two fully rotating inverting prisms in a shielded spectacle frame. The inverting prisms produce a lateral inversion of the rays – the world thus turns "upside down". Besides being a lucid demonstration of the functioning of an inverting prism, experiments involving inverting spectacles encourage a better understanding of the visual process and the functioning of the brain. This also enables a better understanding of the visual perception of babies. Even the apparently simplest things in life (holding and picking up objects, drawing, orientating oneself in a room) become inconceivably difficult.