Ward's® pH Buffer Kit
Supplier: ALDON CORP SE
BB0608-KIT
470233-572EA
65.95
USD
470233-572
Ward's® pH Buffer Kit
Buffers
pH Calibration Buffers
Includes 500 ml each of buffer solution pH 10, pH 4, and pH 7. Also includes 100 ml of electrode storage solution, KCl.
- Aligned with GLOBE protocols
Shelf life: If stored in unopened bottles, then it is one year from receipt. Once opened it should be used within six months of first use.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...