Wallcur® PRACTI-Powders
Supplier: WALLCUR, LLC.
402PP 417MP 425DP 427PW 436VB 469PL 473ZP 480AP 481ML 651AX 710GN 1525DS 496GP
470232-798BX 77.19 USD
470232-798 470232-826 470232-842 470232-846 470232-864 470232-916 470232-926 470232-932 470232-944 470232-946 470233-004 470233-030 470354-902 470355-056
Wallcur® PRACTI-Powders
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
For Reconstitution Practice!
  • Contains No Medication
  • Teaches Proper Reconstitution
  • Can Be Reconstituted With Distilled Water, Or Saline
PRACTI-Powder , for clinical training, is a white powder to simulate multiple drug dosages for reconstitution. Each vial contains a white powder that dissolves in either 0.9% NaCl or water. The PRACTI-Powder vial label is designed to accurately reflect multiple antibiotic dosage strength preparations.
Teach your students these skills:
Adding and mixing diluent to a powder
Calculating and measuring proper dosages
Methods of administration of an oral suspension
Read and interpret medication labels
Sterile vial opening and handling
Air displacement and dosage withdrawal
Needle changes and syringe techniques
Delivery information: These products require a release form for purchase. If your account does not have one on file we can not ship until we receive your release documentation.
Caution: For Training Purposes only, not for Human or Animal use