Practi-Fentanyl™ 5 ml Ampule for Training, Wallcur
Supplier: WALLCUR, LLC.
477FN
470232-940BX 124.29 USD
470232-940
Practi-Fentanyl™ 5 ml Ampule for Training, Wallcur
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
Medication admistration simulation and training.
  • For training purposes only
Practi-Fentanyl™ 5 ml Ampule for clinical training, accurately simulates 250 mcg/5 ml (50 mcg/ml) (0.05 mg/ml) of the synthetic opioid analgesic, Fentanyl. Each of it comes as a labeled 5 ml ampule containing distilled water.
Caution: Not for human or animal use.