TI-Innovator™ Rover Starter Bundle
Supplier: Avantor
470232-632KT 468.59 USD
470232-632
TI-Innovator™ Rover Starter Bundle
Educational Materials Physics Educational Materials Applied Physics Learning Activities
The programmable robotic vehicle that drives conceptual curiosity in math, science and coding.
  • Includes the TI-Innovator Rover, the TI-Innovator Hub and one TI 84 Plus CE calculator
  • Motion enables students to engage in math from a new perspective
  • The simple programming language that is built into TI graphing technology makes it easy to program the system, run it and troubleshoot to correct or fine-tune performance Rover's design allows for access to the TI-Innovator™ Hub input and output ports, so students can add additional capabilities to their Rover experience
Students program Rover to put math and science in motion, adding a physical dimension to verbal, symbolic and graphic representations. Rover also provides an accessible on-ramp to more advanced coding, STEM and robotics projects.
The TI-Innovator™ Hub translates commands from students' TI graphing calculators to control Rover. It can also be used separately from Rover to help students develop basic coding and design skills, and build working STEM and engineering projects.
TI-84 Plus CE graphing calculators open new channels for inquiry, exploration and connection as students use the built-in TI-Basic coding language to program the TI-Innovator™ components.