Somso® Marchantia
Supplier: MARCUS SOMMER
BOS14/3-A
470232-404EA 681.45 USD
470232-404
Somso® Marchantia
Models Botany Models
Somso® premium quality models.
  • 5 year warranty
Marchantia polymorpha, enlarged approx. 10 times, in SOMSO-Plast®. After Prof. Dr. W. Weber. Thallus with three gemma cups each containing gemmae. The thallus becomes either male or female by attaching the two antheridial or two archegonial branches. One antheridial branch has been cut vertically and one part of it can be removed. Separable into 5 parts. On a green base with explanatory note.
Ordering information: Items on this page are not stocked and may have an extended time for delivery.