iworx® 8 Channel Biopotential Amplifier
Supplier: IWORX SYSTEMS INC.
IX-BIO8
470231-650EA 5832.59 USD
470231-650
iworx® 8 Channel Biopotential Amplifier
Data Loggers
An eight Channel Biopotential Amplifier that allows multiple channels of EMG, ECG, EOG to be recorded simultaneously from a single exercising or resting subject. The recorder is furnished with LabScribe Recording and Analysis Software and connects directly to a USB port on either a Macintosh or Windows. The recorder is connected to the subject by snap-leads to pre-gelled Ag/AgCl electrodes.
  • Portable, USB powered
  • Colour coded leads
  • Easy export with Labscribe software
Kinesiology is the study of human and nonhuman animal-body movements, performance, and function by applying the sciences of biomechanics, anatomy, physiology, psychology, and neuroscience. When studying kinesiology, surface electromyography (sEMG or Surface EMG) is the measurement of choice. This recorder enables 8 channels of EMG recordings, or a combination of EMG and ECG recordings. Measuring 2 channels of ECG, Lead I and Lead II allows LabScribe to calculate Lead III, aVR, aVF and aVL . EMG signals can be measured at the same time as the ECG signals.
LabScribe Software System Requirements: Windows: Vista, 7, 7, 10; Mac: OSX,10.9, above;
Min. Requirements: Pentium IV 1GHz, 2GB RAM, 2GB Free Hard drive Space, 2 Available USB ports; Recommended Specs: Quad Core 64 bit processor, 4GB or More Memory"