Ward's® EcoStir
Supplier: VWR International
470231-612
470231-612EA 108.9 USD
470231-612
Ward's® EcoStir
Stirrers Magnetic Stirrers
This lightweight mixing plate is perfect for small laboratory tasks. Weighing in at about two pounds, it's small but mighty.
  • Mixes up to 1.5 L at speeds from 300 to 2000 rpm
  • PET top provides excellent chemical resistance
  • Includes support shaft for use with probe ware
  • Stir bar required but not included
The Ward's® EcoStir has a brushless DC motor that's maintenance free. The ultra silent and vibration-free design allows lab use without disturbing others. The 4.7" workspace is made of PET plastic to help protect against spills.