Innovating Science® Oxidation-Reduction Reactions
Supplier: Ward's Science
IS8013 IS8013-SGL
470163-092KT 51.95 USD
470163-092 470231-560
Innovating Science® Oxidation-Reduction Reactions
Educational Materials Chemistry Educational Materials Microchemistry Learning Activities
Study oxidation-reduction reactions.
  • Classic AP Chemistry Experiment
The term oxidation can mean the chemical combination of a substance with oxygen and reduction can be the removal of oxygen from a compound. When oxygen reacts with any other element (except fluorine) it acquires electrons from that element. The element that donated the electrons is said to be reduced. Three experiments will be run where a compound, which is colorless in solution when reduced, is converted to a deeply colored solution when oxidized. The complete balanced reactions for each step should be written showing the transfer of electrons during oxidation and reduction.
Ordering information: Includes Ferrous Ammonium Sulfate, Sulfuric Acid, Potassium Thiocyanate Solution, Potassium Permanganate, Hydrogen Peroxide, Stannous Chloride Solution, Methylene Blue, Potassium Hydroxide Solution, and Dextrose.
Delivery information: Instructor's Manual and SDS included.