Student Inorganic and Organic Molecular Model Set, 65 Pieces
Supplier: Wards
CH0611
470231-426EA 31.55 USD
470231-426
Student Inorganic and Organic Molecular Model Set, 65 Pieces
Models Molecular Biology Models
Perfect Starter Set
  • 32 atoms
  • 33 bonds
Contents: 6 Black Carbon (4 Hole, 22 mm), 10 White Hydrogen (1 Hole, 15 mm), 4 Red Oxygen (2 Hole, 20 mm), 2 Blue Nitrogen (4 Hole, 22 mm), 2 Green Halogen (1 Hole, 20 mm), 2 Green Halogen (4 Hole, 22 mm), 1 Yellow Sulfur (2 Hole, 22 mm), 1 Yellow Sulfur (4 hole, 22 mm), 1 Purple Phosphorus (4 Hole, 22 mm), 2 Grey Metal (1 Hole, 15 mm), 1 Grey Metal (2 Hole, 22 mm), 8 Long Grey Bonds, 25 medium Grey Bonds.