Ward's® Essentials Crawling Color Walking Water
Supplier: Science First
670-1100
470231-282KT 56.09 USD
470231-282
Ward's® Essentials Crawling Color Walking Water
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
Explore properties of light with primary and secondary colors.
  • Durable acrylic display case for capillary action demonstration
  • Student-directed inquiry color mixing experiments
  • Designed for chemistry or biology classrooms
  • Aligned with NGSS MS-PS4, MS-LS1
This high-quality acrylic display allows the classroom to explore color mixing and the properties of water, including cohesion, adhesion, and capillary action. Students take their knowledge a step further by engineering a plant model and mapping capillary action through the 'xylem.' This kit includes enough materials for 15 student groups.
Ordering information: Kit includes acrylic setup, food coloring, mixing sheets, disposable pipets, and teacher’s guide.