Stainless Steel Test Tube Rack
Supplier: United Scientific Supplies
TTRSS25
470231-002EA 21.25 USD
470231-002
Stainless Steel Test Tube Rack
Racks Tube Racks
Convenient storage for tubes.
  • Stainless Steel
  • Holds 25 tubes up to 16 mm in diameter
  • Easy cleaning and labeling
Sturdy Z-shaped rack made from a single sheet of heavy gauge 304 stainless steel.