3B Scientific® Cell Anatomy Models Bundle
Supplier: Avantor
470230-928EA 1320.49 USD
470230-928
3B Scientific® Cell Anatomy Models Bundle
Models Molecular Biology Models
Modeled After EM Scans.
  • Materials resist breakage
  • Buy as a set and save
  • 3-year manufacturers warranty
These two cell models show the anatomy of plant and animal cells in very high detail. Not just a basic conceptual model, but constructed from scans using electron microscopy. Organelles are presented in a raised relief from the surface, and structures are identified with included keys. Even the removable bases on these models are functional, showing the cells placement with adjacent cells and even showing the plasmodesmata between the plant cells.