Polymers - Making Fluorescent Worms
Supplier: Ward's Science
IS7045
470230-900EA 54.95 USD
470230-900
Polymers - Making Fluorescent Worms
Educational Materials Chemistry Educational Materials Applied Chemistry Learning Activities
Create your own fluorescent worms while understanding polymer chemistry and cross-linking.
  • Understand and demonstrate polymer chemistry and cross-linking htorugh a synyhesis reaction
  • Aligned to NGSS practices
  • Cross cutting concepts: patterns
  • Disciplinary core ideas: PS1.A, PS1.B
  • Learn engineering practices: analyzing and interpreting data
Allow students to learn about polymers and cross-linking using this fun and colorful activity. Students add a solution of sodium alginate to three calcium chloride solutions, each containing a different fluorescent dye. After a few minutes the liquid sodium alginate solution forms into gel-like polymer 'worms'. Upon completion of the activity, the worms may be placed under a black light and the students will observe their worms glow red, blue, and green.
Caution: WARNING: This product can expose you to chemicals including D&C Red no. 19, which are known to the state of California to cause cancer. For more information go to www.P65Warnings.ca.gov