Ward's® Mini Digital Incubator
Supplier: VWR International
470230-608
470230-608EA 806 USD
470230-608
Ward's® Mini Digital Incubator
Incubators Standard Incubators
Personal Mini Incubator Perfect for Every Lab.
  • 20 liter capacity
  • Digital temperature control
  • Personal sized, 13 x 14.5" footprint
  • Accepts bottles and flasks up to 2L
  • Internal power outlet, for installation of small shakers, mixers or other instruments
With digital temperature control, the Ward's Mini Digital Incubator provides convenient "set and walk away" operation, eliminating the need for external thermometers and repetitive "fine tuning" of an analog control knob. Simply choose the desired temperature and the incubator immediately begins to heat up while accurately monitoring the chamber temperature and conveniently displaying the temperature in real time on the large LED control panel.
Despite a modest footprint, this incubator features large internal chambers, capable of storing flasks and bottles up to 2 liters. In addition, this incubator includes 2 removable shelves and a built-in (universal) internal outlet for powering small shakers, rockers, etc.