Neulog Thermal Conductivity Kit
Supplier: Wards
THRKIT
470230-452KT 45.3 USD
470230-452
Neulog Thermal Conductivity Kit
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Ideal for study of thermal conductivity.
  • Study thermal conductivity of various solids
This kit allows for a qualitative and quantitative study of the thermal conductivity of different solids such as copper, brass and steel. NeuLog Temperature Sensor ( Labs Part Number 233) NOT INCLUDED.
Ordering information: NeuLog Temperature Sensor ( Labs Part Number 233) NOT INCLUDED.
Caution: For educational use only.