St. Louis Motor Basic
Supplier: Wards
PH1240N8
470230-434EA 56.85 USD
470230-434
St. Louis Motor Basic
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Demonstrates operation of motors and generators.
  • St Louis motor demonstration is useful in demonstrating the operations of motor and generator
  • Consisting of a two-pole armature mounted between magnets
  • Steel supports are mounted on a ridged 4" dia. plastic base
  • Will operate on 3 to 6 V DC
  • Power supply not included
St Louis Motor Demonstration is useful in demonstrating the operations of motor and generator. Consisting of a two-pole armature mounted between magnets. Steel supports are mounted on a ridged 4" Diameter plastic base. Will operate on 3 to 6 V DC. Power supply not included. This open design allows students to see each part of the motor operate, which shows basic function.