Hand Held Centripetal Force Kit

Supplier: Wards
PH1120152
470230-402KT 11.85 USD
470230-402
Hand Held Centripetal Force Kit
Educational Materials Physics Educational Materials Mechanics Learning Activities
Ideal for studying rotational dynamics.

  • Kit Contains: (1) Hollow plastic grip handle, (1) String, (20) Washers, (2) Paper Clips, (1) Rubber stopper
  • Student puts string through handle, spins rubber stopper around and balances that with the masses at the bottom of the string
  • Activity allows students to study how mass and radius affect centripetal acceleration and rotational velocity
  • Easy fund demonstration of centripetal force

This elegantly simple kit provides the necessary tools to discover properties of rotational dynamics. A rubber stopper swings around on the end of a string counter balanced by weights, allowing students to see how mass and radius affect centripetal acceleration and rotational velocity.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR