Belts for Wimshurst
Supplier: Wards
PH0848BELT
470230-304PK 20.1 USD
470230-304
Belts for Wimshurst
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Spare belt for Wimshurst machine. This item has two replacement belts, both of which have a 20" circumference.
  • Smaller dia. belt dia. 0.13"
  • Larger dia. belt dia. 0.20"
  • Black in color
  • Flexible rubber material
The flexible rubber belts is for use with the Wimshurst Machine (SKU# PH0848C or SKU# PH0848D) - this machine (with the belts) is a hand-cranked electrostatic generator that produces voltage and, in essence, creates lightening!