Magnetizing Charger
Supplier: Wards
PH0800E
470230-266EA 83.7 USD
470230-266
Magnetizing Charger
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Magnetizing box for recharging magnets.
  • Recharge your magnet with out power
  • 8 x 5" base
  • Charger for bar and horseshoe magnets
  • No plugs and no batteries needed
Labs Magnetizing Box 8" x 5"- Recharge your Magnet with out Power - Charger for Bar and Horseshoe magnets