Vibration Generator

Supplier: Wards
PH0769A
470230-260EA 97.1 USD
470230-260
Vibration Generator
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Provides electronically controlled waves.

  • Product is used for generating waves with a string or for use with a ripple tank
  • Overall height: 5.5", overall diameter: 4.5"
  • Plastic item with foam inside
  • For use with square wave generator

The vibration generator is a sturdy, user-friendly tool for providing electronically controlled waves - this allows one to create ripples for the study of wave motion (recommended for use with a square wave generator).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR