Steel Tuning Forks, Set of 8
Supplier: Wards
PH0738ALT
470230-246EA 94.4 USD
470230-246
Steel Tuning Forks, Set of 8
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Steel tuning forks for physics experiments.
  • ±5% of stated frequency
Steel Tuning Forks, Set of 8, C (256), D (288), E (320), F (341), G (384), A (426), B (480), C (512).