Steel Tuning Forks with Wood Case
Supplier: Wards
PH0738A
470230-244EA 55.6 USD
470230-244
Steel Tuning Forks with Wood Case
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Steel tuning forks for physics experiments.
  • Supplied in wooden case
Nickel-plated, plain shanks, with frequency marked, C (256), D (288), E (320), F (341), G (384), A (426), B (480), C (512). Set supplied in wooden case.