Wave Apparatus, Plastic
Supplier: Wards
PH0712N8
470230-242EA 115.2 USD
470230-242
Wave Apparatus, Plastic
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Plastic wave apparatus ideal for studying wave behavior.
  • Demonstrate longitudinal and transverse waves
This advanced apparatus demonstrates both longitudinal and transverse wave motions. Consisting of a number of eccentric discs supporting a series of metal rods revolving on a handle, transverse waves are obtained. Longitudinal waves are obtained from the bent rods.