Wave Form Helix, 3m
Supplier: Wards
PH0706
470230-234EA 37.9 USD
470230-234
Wave Form Helix, 3m
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Closely wound steel helix for studying wave behavior.
  • Closely wound helix comprised of steel wire
  • 20 mm dia.
  • Closed length of 3 meters
  • Extends to 9 meters
  • Supplied with manual
Steel wire close-wounded helix 20 mm diameter, closed length 3 meters extending to approx. 9 meters extended, with looped ends.