Power Pack
Supplier: Wards
PH0676A
470230-232EA 137.55 USD
470230-232
Power Pack
Educational Materials Physics Educational Materials Optics Learning Activities
Low voltage power supply that can be used as a battery eliminator.
  • Switchable between 6 V or 12 V, AC or DC (at 2 Amps, not adjustable)
  • Banana plug terminals
  • Perfect for use with optical benches or other physics equipment
  • Can be used as a high powered battery eliminator
Power pack is a low voltage power supply that operates on 110 V power and outputs 6 or 12 V at 2 Amp either AC or DC.