Spectrometer Basic
Supplier: Wards
PH0618A
470230-218EA 243.75 USD
470230-218
Spectrometer Basic
Educational Materials Physics Educational Materials Optics Learning Activities
Spectrometer for studying basic concepts of optics.
  • Basic Spectrometer
  • Adjustable collimator tube, telescope, and prism stand
  • Approximately 9 inches in height
  • Sturdy metal base
  • For studying basic concepts of optics (physics of light)
The basic spectrometer, with adjustable collimator tube, telescope, and prism stand, is a sturdy tool with which to study various basic concepts of optics (physics of light) (reflection and diffraction, for example) in a visually and kinesthetically effective manner.