Eisco® Convection of Gas Apparatus, Small
Supplier: Wards
PH0410B
470230-170EA 44.6 USD
470230-170
Eisco® Convection of Gas Apparatus, Small
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Apparatus to study convection of gas.
  • Smaller sized to fit in any classroom than standard convection of gas apparatus
  • Candles not included
Place a small candle under one chimney to heat the air inside the chamber. A convection current be begin moving, drawing fresh, colder air in through the other chimney. Use a piece of smoking paper to vividly demonstrate the movement of air through the cell.
Accessories information:
This apparatus is comprised of a 10×4×3" metal box with a sliding front door, two glass chimneys and a candle holder. Candles and paper are not included.