Rods for Thermal Conductivity

Supplier: Wards
PH0397G
470230-168EA 19.55 USD
470230-168
Rods for Thermal Conductivity
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Various rods to explore thermal conductivity.

  • Contains iron, copper, aluminum, lead, brass, and zinc rods
  • Used for thermal conductivity experiments

The Thermal Conductivity Rods provide an excellent tool for use in thermal conductivity experiments. Rods made with 6 different metals are included (Iron, Copper, Aluminum, Lead, Brass, and Zinc), making studying thermal conductivity of various metals
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR