Bimetallic Strips on Blocks, Set 10
Supplier: Wards
PH0382
470230-164PK 41.95 USD
470230-164
Bimetallic Strips on Blocks, Set 10
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Bimetallic strips attached to wood blocks to study thermal energy.
  • 10 strips per pack
  • Bimetallic strip attached to wood block (block 1.85 x 1.45 x 0.4")
  • Total length of item: 4.31"
  • Strips attached by 2 nails
Bimetallic strips are sturdy strips of two metals attached to solid wood blocks and can be used to study thermal energy converting to mechanical motion (this is caused by the different expansions of two different metals when heated).