Bar Breaking Apparatus

Supplier: Wards
PH0380A
470230-162EA 85.5 USD
470230-162
Bar Breaking Apparatus
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
This apparatus is useful for studying forces during thermal contraction and expansion.

  • 13" long bar
  • Contains iron base, bar with tension nut, and breaking bars

The bar breaking apparatus provides a very sturdy and user-friendly method by which to introduce students to the forces that can occur during thermal contraction and thermal expansion. Students can experience and study the forces of thermal contraction.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR