Inclined Plane Steel with Pulley
Supplier: Wards
PH0335
470230-124EA 85.45 USD
470230-124
Inclined Plane Steel with Pulley
Educational Materials Physics Educational Materials Mechanics Learning Activities
Steel inclined plane with accessories.
  • 48” long and straight as an arrow.
  • Made of heavy steel with rounded corners
  • Includes car, clamp pulley, string and basket
  • Stand base, rod and support clamp are NOT included
This green inclined plane is 48" long and straight as an arrow. It is made of heavy steel with rounded corners and can accommodate any kind of car. Holes in the supporting flanges fit a 12 mm support rod. Includes car ,clamp pulley, string and basket. Stand base, rod and support clamp are not included.