Spouting Cylinder
Supplier: Wards
PH0196
470230-064EA 31.55 USD
470230-064
Spouting Cylinder
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
Durable shatter-proof plastic.
  • Demonstrating principles of Snell's Law
  • 2.3" (60 mm) dia. pipe
  • 15.5" (400 mm) total height
Demonstrate Snell's Law from an acrylic pipe. The pipe is 15.5" tall and 2.3" in diameter and has holes at predetermined heights.