Bourdon Gauge
Supplier: Wards
PH0139
470230-056EA 86.9 USD
470230-056
Bourdon Gauge
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
A working gauge is mounted in a transparent case so the actual working parts are visible.
  • Black base with gauge and nipple (0.340in dia.)
  • Gauge enclosed in clear plastic case to allow inner workings to be visible
  • Kg/cm2 delineated with black markings and psi delineated with red markings
The Bourdon gauge is a safe and simple tool for measuring the pressure of various gases and liquids. The stand has a nipple with which to connect a gas or liquid source hose and its easy to read gauge makes it very user-friendly.