Density, Set Assorted Shapes, Set 14

Supplier: Wards
PH0114A
470230-054EA 145.1 USD
470230-054
Density, Set Assorted Shapes, Set 14
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
Fourteen materials in the form of rectangular blocks.

  • 14 different blocks
  • Kit contains 11 different materials,
  • This kit is intended to familiarize students with appearance, feel, texture and density of these common materials.
  • All measurements are approx. and may vary from kit to kit

Ordering information: Kit consists of 14 blocks: Aluminum Block (2" x 1.5" x 1.125"); Aluminum Rectangle (3.875" x 0.75" x 0.75"); Brass Block (2" x 0.75" x 0.75"); Styrofoam block (2" x 1.5" x 1.125"); Wax Block (2" x 1.5" 1.125"); Stainless Steel Block (2" x 2" x 0.75”); Wood Block (2" x 1.5625" x 1.1875”); Wood Rectangle (3.875" x 0.75" x 0.75"); Soap stone Rectangle (3.875" x 0.75" x 0.75"); Marble Rectangle (3.875" x 0.75" x 0.75"); Glass Rectangle (3.875" x 0.75" x 0.75"); Acrylic Rectangle (3.875" x 0.75" x 0.75"); Steel Block (1.5" x 1.5" x 0.75"); Steel Block (2" x 1.5" x 1.125").
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR