Density Super Pack

Supplier: Wards
PH0113SET
470230-052EA 266.65 USD
470230-052
Density Super Pack
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
4 Holders, 12 Al Rods, 12 Brass Rods, 12 PVC Rods, 12 Polypropylene Rods

  • (12) Aluminum Rods, (12) Brass Rods, (12) Polypropylene Rods, (12) PVC Rods, 4 Bases to hold all rods
  • Rods vary in length from 1.5" to 2.625" per material set
  • To determine the density of the material the student plots mass versus volume and the slope therefore is the density (mass per unit volume). Students will need to do all measurements themselves so a balance and ruler are required
  • Bases are made from a durable wood coated for years of use
  • Machined from top quality raw materials to produce accurate results and durability of decades of use

The perfect demonstrational tool to show that the density of a substance is constant regardless of mass and volume. Using this kit and entire classroom of students can working measuring and calculating densities of the different materials and volumes. (12) Aluminum Rods, (12) Brass Rods, (12) Polypropylene Rods, (12) PVC Rods, 4 Bases to hold all rods
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR