Equal Mass, Set 5 cylinders
Supplier: Wards
PH0112FCB
470230-042EA 30.05 USD
470230-042
Equal Mass, Set 5 cylinders
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
To study the relationship between mass and density.
  • 5 Cylinders, 0.5 inch dia..
  • Aluminum, Copper, Tin, Zinc, Lead
To study the relationship between mass and density. Set of 5 cylinders having same mass and diameter 0.50" varying lengths.