Specific Gravity Cylinders

Supplier: Wards
PH0110
470230-040EA 20.15 USD
470230-040
Specific Gravity Cylinders
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
Four different metal cylinders each measuring 38 mm long x 10 mm in diameter.

  • 4 metal cylinders for the study of gravity, density, and mass
  • Aluminum, zinc, copper and steel
  • Comes with hook holes and removable hooks to be used in a variety of experiments

Four different metal cylinders each measuring 38 mm long x 10 mm in diameter, made of aluminum, zinc, copper and steel. Comes with hook holes and removable hooks to be used in a variety of experiments.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR