Density Cubes with Hooks

Supplier: Wards
PH0108PCB
470230-032EA 62.2 USD
470230-032
Density Cubes with Hooks
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
Density Cubes, Set of 4 Metals with Hooks, 1.26" (32 mm) sides.

  • Brass, lead, zinc, copper, aluminum, and iron cubes
  • 32 mm cubes with hooks

The set consists of four 32mm cubes of the following metals: aluminum, brass, lead, and steel . Perfect for investigating density. Note: this product contains lead. To be used for scientific purposes only.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR