Specific Gravity Cubes

Supplier: Wards
PH0108 PH0108A PH0108D PH0108E PH0108F
470230-020EA 31.35 USD
470230-020 470230-022 470230-024 470230-026 470230-028
Specific Gravity Cubes
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
Specific Gravity Cubes with Hooks.

  • Brass, iron, aluminum and zinc 20mm cubes
  • Hooks

Specific Gravity Cubes with Hooks (Set of 4) This density cube set contains the follow metal cubes: brass, iron, aluminum and zinc. They are .79in (20mm) cubes.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR