Micrometer with Lock, Eisco Scientific
Supplier: Wards
PH0086B
470230-014EA 18.3 USD
470230-014
Micrometer with Lock, Eisco Scientific
Calipers
Made of nickel plated brass, with ratchet top, accurately machined stainless steel rod.
Micrometer Screw Gauge w/Lock, Nickel Plated Brass - Range 0-25 x 0.01 mm, supplied in case. Accuracy is perfect for a hobbyist or student -- at 10mm, accuracy is +/- .003mm.