Micrometer, Eisco Scientific
Supplier: Wards
PH0086A
470230-012EA 18.65 USD
470230-012
Micrometer, Eisco Scientific
Calipers
Made of nickel plated brass, with ratchet top, accurately machined stainless steel rod.
Micrometer Screw Gauge, Nickel Plated Brass - Range 0-25 x 0.01 mm (1) Micrometer. Range 0-25 x 0.01 mm, supplied in case. Accuracy is perfect for a hobbyist or student -- at 10 mm, accuracy is ± .003 mm.