USB B/M to B/M Cable, Eisco Scientific
Supplier: Wards
USBBBMALE
470229-986EA 20.6 USD
470229-986
USB B/M to B/M Cable, Eisco Scientific
Cables
1m Neulog B/M to B/M Cable.
  • USB 2.0 converter cable takes Type B Male connection to Type B Male connection
  • 1 meter in length
USB 2.0 Cable - Type B Male to Type B Male - Converter Cable, 1m. Standard USB 2.0 B/M to B/M cable.