Neulog Solar Cell Kit
Supplier: Wards
SLSES
470229-982KT 38 USD
470229-982
Neulog Solar Cell Kit
Data Loggers
Hardware to conduct solar power experiments with NeuLog sensors.
  • Kit enables students to study: Photovoltaic cells, light sources, color sensitivity of photovoltaic cells
Uses different light sources (incandescent/fluorescent/sunlight) and color filters to investigate the amount of energy irradiated from a light source, as well as the sensitivity of the solar cell as a function of the color (frequency) of irradiation.
Ordering information: Includes Solar cell, three color filters, flashlight, instructions.
Caution: For educational use only.