Neulog Inclined Plane Kit
Supplier: Wards
INPSES
470229-970KT 115.6 USD
470229-970
Neulog Inclined Plane Kit
Educational Classroom Kits and Activities
Complete inclined plane kit with cart and pulley.
  • Steel painted incline plane - 18 inches of runway
  • Cart has low friction bearings
  • Great for use with a photogate due to vertical rod
  • Great for use with motion sensor due to sail attachment
Complete incline plane kit with 18" incline runway, cart, friction blocks, pulley, string, measuring devices
Ordering information: Kit includes (1) Incline plane with base, (1) Hinge hardware, (1) Measurement Arch, (1) Cart with vertical rod, (1) Sail for cart, (1) Tape Measure, 4+ Meters of nylon string, (1) pulley with extension rod and mounting hardware, (1) Solid Friction Block, (1) Open Friction Block.
Caution: For educational use only.