Gas Law Demonstration
Supplier: Wards
GSLWSES
470229-968EA 109.95 USD
470229-968
Gas Law Demonstration
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
Investigate the dependence of pressure on a gas with changes in temperature and volume.
  • Gas Law apparatus with 55ml gas volume
  • 50ml syringe
  • Rubber stopper
  • Three tea candles
  • Instructions
Investigate the dependence of pressure on a gas with changes in temperature and volume.