Neodymium Sphere Magnet
Supplier: Master Magnetics
5XNS50BX
470229-352EA 16.55 USD
470229-352
Neodymium Sphere Magnet
Magnets
A round neodymium magnet for exciting magnet inquiry.
  • Very high resistance to demagnetization
  • High energy for size
  • Good in ambient temperature
Neodymium Disk Magnets (Nd-Fe-B) are made from the most powerful magnetic material in the world. They boast extreme strength in a small size. The sphere magnet has a north and south pole, one on each half of the sphere. It has a grade of 35 with a Gauss rating of 12,200. Maximum operating temperature is 80°C (176°F).