Stem Explorations, Gears Building Set
Supplier: K'nex Industries
79318
470229-232CS 107.2 USD
470229-232
Stem Explorations, Gears Building Set
Educational Materials Physics Educational Materials Mechanics Learning Activities
Explore STEM concepts while building different geared models!
  • Students will learn about gear configurations, gear ratios, rotational speed, mechanical advantage and more!
Using the materials included in this set, elementary-school aged children will be engaged and energized as they further their knowledge and understanding of science, technology, engineering and math concepts. The set includes 140+ parts for a single child or team of 2 children to build 3 unique geared models: a Crank Fan, a Car Window and an Egg Beater. Once built, a downloadable booklet guides students through 3 hands-on, inquiry-based experiments on each of the models.