TeacherGeek Soldering Package
Supplier: TEACHERGEEK INC
1834-53
470229-114KT 30 USD
470229-114
TeacherGeek Soldering Package
Educational Materials Physics Educational Materials TeacherGeek System Components
Here's everything you will need to get soldering.
  • A high quality, ceramic element soldering pencil
  • 20 g (6 ft.) of lead-free solder in a pencil dispenser
  • A mini soldering iron stand